missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma mPAGE™ 4X LDS Sample Buffer

Catalog No. 11580587
Change view
Click to view available options
Buffer:
4X LDS Sample Buffer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buffer
11580587 4X LDS Sample Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11580587 Supplier MilliporeSigma Supplier No. MPSB250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to prepare protein samples for denaturing polyacrylamide gel electrophoresis (PAGE) with mPAGE and most other Bis-Tris gels.

  • Used to prepare protein samples for denaturing polyacrylamide gel electrophoresis (PAGE) with mPAGE and most other Bis-Tris gels. LDS sample buffer contains lithium dodecyl sulfate with pH at 8.4 for optimal activity of the reducing agent.
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Specifications

Buffer 4X LDS Sample Buffer
Concentration 4X LDS Sample Buffer
pH Range 8.4
Separation of Protein
Quantity 1 Ea.
Product Type LDS sample buffer