missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma mPAGE™ 4-20% Bis-Tris Precast Gel

Precast gel for research use only

$173.00 - $174.00

Specifications

Buffer MES or MOPS Running Buffer
Concentration 4 to 20% Bis to Tris
For Use With (Equipment) Mini Gel Systems
Gel Percentage 4 to 20%
GelThickness 1 mm
View More Specs

Products 3
Catalog Number Mfr. No. Wells Price Quantity  
Catalog Number Mfr. No. Wells Price Quantity  
11-580-609
View Documents
MilliporeSigma
MP42G10
10-well
Pack of 10 Each for $173.00
Only null left
 
11-580-608
View Documents
MilliporeSigma
MP42G12
12-well
Pack of 10 Each for $174.00
Only null left
 
11-580-583
View Documents
MilliporeSigma
MP42G15
15-well
Pack of 10 Each for $174.00
Only null left
 
Description

Description

  • mPAGE 4-20% Bis-Tris Precast Gel, 10x8 cm
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications

Specifications

MES or MOPS Running Buffer
Mini Gel Systems
1 mm
Bis-Tris precast gels
10 Pk.
8 cm
4 to 20% Bis to Tris
4 to 20%
Bis-Tris
Protein
10 cm
Videos
SDS
Documents

Documents