missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma mPAGE™ 4-12% Bis-Tris Precast Gel

Catalog No. 11580591
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
11580591 15-well
11580601 10-well
11580590 12-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11580591 Supplier MilliporeSigma Supplier No. MP41G15
Only null left

Precast gel for research use only

  • mPAGE 4-12% Bis-Tris Precast Gel, 10x8 cm
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Specifications

Buffer MES or MOPS Running Buffer
Concentration 4 to 12% Bis to Tris
Gel Percentage 4 to 12%
Gel Thickness 1 mm
Gel Type Bis-Tris
Sample Loading Volume 40 μL
Separation of Protein
Wells 15-well
Quantity 10 Pk.
Length (Metric) 10 cm
Width (Metric) 8 cm
Product Type Bis-Tris Precast Gels
For Use With (Equipment) Mini Gel Systems
Show More Show Less