missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma mPAGE™ 12% Bis-Tris Precast Gel

Precast gel for research use only

Supplier:  MilliporeSigma MP12W12

 View more versions of this product

Catalog No. 11-580-595


Only null left
Explore available promotions

Description

Description

  • mPAGE 12% Bis-Tris Precast Gel, 10x8 cm
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications

Specifications

MES or MOPS Running Buffer
Mini Gel Systems
1 mm
Bis-Tris Precast Gels
Protein
10 Pk.
8 cm
12% Bis-Tris
12%
Bis-Tris
60 μL
12-well
10 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions