missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast Hydrolysate Enzymatic

Autolyzed Brewers yeast

$103.15 - $1341.39

Specifications

Content And Storage 15-30°C
Form Tan powder
Source Brewers Yeast
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10330405
View Documents
MP Biomedicals™
0210330405
5 lb.
Each for $302.72
Only null left
 
ICN10330425
View Documents
MP Biomedicals™
0210330425
25 lb.
Each for $1,341.39
Only null left
 
ICN10330401
View Documents
MP Biomedicals™
0210330401
1 lb.
Each for $103.15
Only null left
 
Description

Description

Yeast Hydrolysate Enzymatic is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.

Specifications

Specifications

15-30°C
Tan powder
Brewers Yeast
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'