Learn More
MP Biomedicals™ Yeast Extract Powder
A vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of approximately three times the weight of ordinary dry yeast.
Supplier: MP Biomedicals™ 0210330390
Specifications
| 15° to 30°C | |
| Yeast | |
| 500 g |
| Powder | |
| 5 to 5.5 |