missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 0.05% Trypsin-EDTA (1X)

Catalog No. ICN1689149
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1689149 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1689149 Supplier MP Biomedicals™ Supplier No. 091689149

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

0.05% Trypsin and 0.5mM EDTA in modified HBSS w/o Ca++ and Mg++, 100 mL

Concentration: 0.05% (w/v) Trypsin (1:250) and 0.02% (w/v) EDTA (Versene)

Specifications

For Use With (Application) Cell dissociation, routine cell culture passaging and primary tissue dissociation due to its digestive strength
pH Range 7 to 7.4
Sample Type Porcine
Content And Storage 0°C to 20°C
Form Solution
Quantity 100 mL
Product Type Trypsin-EDTA