missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Trypsin, 1 - 250, From Hog Pancreas
From Hog Pancreas
Supplier: MP Biomedicals™ 0210313980
Description
Trypsin, 1 - 250, [9002-07-7]
- Parenzymol
- Dry powder: stable
- Protease act: min 250,000 USP units/gm
Dry Powder
Specifications
| 24000 | |
| ∽8 | |
| Hog | |
| Powder |
| Trypsin | |
| 100 g | |
| 4°C |