missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Trypsin, 1 - 250, From Hog Pancreas

Catalog No. p-3179315
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10313925 25 g
ICN10313980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10313925 Supplier MP Biomedicals™ Supplier No. 0210313925
Only null left

From Hog Pancreas

Trypsin, 1 - 250, [9002-07-7]

  • Parenzymol
  • Dry powder: stable
  • Protease act: min 250,000 USP units/gm

Dry Powder

Specifications

Molecular Weight (g/mol) 24000
Name Trypsin
pH Range ∽8
Quantity 25 g
Source Hog
Storage Requirements 4°C
Form Powder