missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Superoxide Dismutase
From bovine erthrocytes
Supplier: MP Biomedicals™ 0219011730
Description
Activity: 3500U/mg. Unit Definition: That amount of enzyme causing a 50% inhibition in the rate of assay based on the method of McCord, J.M. and Fridovich, J. Biol. Chem., 1: 6049, (1969). Lyophilized salt-free powder.
Specifications
| ≥0.900 mg protein/mg solid | |
| Superoxide Dismutase plays a critical role in the defense of cells against the toxic effects of Oxygen radicals | |
| Bovine erythrocytes | |
| Superoxide Dismutase | |
| Lyophilized Powder |
| Pale Blue | |
| -20°C | |
| 30 kU | |
| ≈32500 |