missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sodium Phosphate Buffer

Catalog No. 116560205
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
116560205 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 116560205 Supplier MP Biomedicals™ Supplier No. 116560205
Only null left

Sodium Phosphate Buffer is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography. It occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4).

  • Component of the FastDNA SPIN Kit for Soil and FastDNA SPIN Kit for Feces.
  • Sodium phosphate is a reagent with very high buffering capacity that is widely used in molecular biology, biochemistry, and chromatography.
  • It occurs in several forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4).

Specifications

Content And Storage Store at Room Temperature (15°C to 30°C)
For Use With (Application) Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
pH 7.8 to 8.2
Product Type Sodium Phosphate Buffer
Quantity 60 mL

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.