missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RNAPro Solution

Catalog No. MP116055050
Change view
Click to view available options
Quantity:
2 x 27.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116055050 2 x 27.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116055050 Supplier MP Biomedicals™ Supplier No. 116055050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RNAPro Solution is a proprietary lysis buffer included with all of the FastRNA Pro purification kits. It can be used with the kits or with any other purification protocol as the primary lysis buffer to isolate total RNA from any type of plant or animal tissue, cultured cells, microbes, or soil and environmental samples.

  • Extracted RNA is suitable for RT-PCR analysis and other downstream applications
  • Using the FastPrep-24™ or FastPrep™ FP120 Instrument, samples can be homogenized by using MP Lysing Matrix tubes
  • Use with FastPrep-24 or FastPrep-96 (with ConeFlex Legacy Adapter), or with optional CoolPrep Adapter kits to lyse, grind or homogenize samples, for the extraction and purification of RNA

Specifications

Content And Storage Room Temperature
Isolation Technology Organic Extraction
Sample Type Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
For Use With (Application) Isolation of RNA from most biological or environmental samples
Product Type RNAPro Solution
Quantity 2 x 27.5 mL
Sample Volume Scalable

Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.