missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ribonuclease A

From bovine pancreas

Supplier:  MP Biomedicals™ 0219398083

 View more versions of this product

Catalog No. ICN19398083


Only null left
Explore available promotions

Description

Description

Chromatographically purified. Salt-free and protease-free. Activity: ≥70 Kunitz units/mg. Unit Definition: One unit causes the hydrolysis of RNA at a rate such that the velocity constant (k) equals 1 at 25°C and pH 5.0. Lyophilized powder.

Specifications

Specifications

≥70 kU/mg
>70%
purification of DNA, analysis of RNA sequences, RNase protection assays, hydrolyze RNA contained in Protein samples
Bovine pancreas
Ribonuclease A
Heavy Metal Ions, DNA
White
7 to 7.5
0°C
250 mg
13700
Lyophilized Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions