missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: ≥70,000 MW
Facilitate attachment, adherence, and spreading in the culture of adherent cells
$113.86 - $113.86
Specifications
| Content And Storage | -20°C |
|---|---|
| Product Type | Poly-L-Lysine |
| Source | Synthetic |
| Catalog Number | Mfr. No. | Quantity | Sterility | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Sterility | Price | Quantity | ||||
|
ICN19454405
|
MP Biomedicals™
0219454405 |
5 mg | Sterile |
Each for $113.86
|
|
||||
Description
Available nonsterile or gamma-irradiated.
Specifications
| -20°C | |
| Poly-L-Lysine | |
| Synthetic |