Learn More
MP Biomedicals™ PlantCon™ Plant Tissue Culture Container
Sterilized Base for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
Manufacturer: MP Biomedicals™ 092672206
Description
Clear polyvinyl chloride (PVC) dishes specifically designed for plant culture applications. Ideal for use in commercial horticulture or research settings. Allow exceptional light transmission and optimal growth. Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization.
Order Info
Base L × W × H: 3 3/4 × 3 3/4 × 1 3/8 in. (9.5 × 9.5 × 3.5cm). Covers and bases packaged separately for shipping purposes. Covers and bases also sold separately. Sterile.
Specifications
Covers Sold Separately (See 092672102) | |
Clear | |
PVC | |
Sterile | |
200/Cs | |
9.5 mm | |
7 mm |
Square | |
Disposable | |
Rectangular | |
Cell Culture Dish | |
3.75 x 3.75 in. (9.5 x 9.5 cm) | |
9.5 mm |