missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ PlantCon™ Plant Tissue Culture Container

Catalog No. p-2625663
Change view
Click to view available options
Quantity:
1 Ea
200/Cs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2672002 1 Ea
ICN2672206 200/Cs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN2672002 Supplier MP Biomedicals™ Supplier No. 092672002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sterilized Base for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.

Clear polyvinyl chloride (PVC) dishes specifically designed for plant culture applications. Ideal for use in commercial horticulture or research settings. Allow exceptional light transmission and optimal growth. Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization.

Order Info

Base L x W x H: 3 3/4 x 3 3/4 x 1 3/8 in. (9.5 x 9.5 x 3.5cm). Covers and bases are sold separately. Sterile.

Specifications

Closure Type Covers Sold Separately (See 092672102)
Bottom Shape Square
Color Clear
Disposable Single-use
Material PVC
Shape Rectangular
Sterility Sterile
Type Cell Culture Dish
Quantity 1 Ea
Dimensions (L x W) 3.75 x 3.75 in. (9.5 x 9.5 cm)
Length (Metric) 9.5 mm
Width (Metric) 9.5 mm
Height (Metric) 7 mm
Show More Show Less