missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Peroxidase, 250-330U/mg solid
From horseradish
Supplier: MP Biomedicals™ 0219537305
Description
Activity: 250-330U/mg solid. Unit Definition: One unit will form 1mg purpurogallin in 20 seconds from pyrogallol at pH 6.0 and 20°C. Essentially salt-free powder.
Specifications
| ≥250 U/mg | |
| 7 | |
| 0°C | |
| 5 kU | |
| ≈44000 | |
| Crystalline Powder |
| Reddish Brown | |
| ELISA, immunohistochemistry, western blotting | |
| Horseradish | |
| Peroxidase | |
| HRP is reversibly inhibited by Cyanide and Sulfide at a concentration of 10-5M, p-Aminobenzoic Acid, Azide, Cyclopropanone, L-cystine, Dichromate, Ethylenethiourea, Hydroxylamine, Sulfite and Vanadate and divalent Metals Cd, Co, Cu, Fe, Mn, Ni, Pb |