missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Papain
Description
Sulfhydryl protease from Carica papaya latex. (A second protease, chymopapain, and a lysozyme have also been isolated from this same source.)
- Format: Powder
- Grade: Technical
- Molecular weight: 23,000
- Specific Activity:>1750 USP Units/mg
Specifications
Specifications
| Color | Off-White |
| pH | 6 to 7 |
| For Use With (Application) | Enzymatic synthesis of Amino Acids, Peptides and other molecules |
| Content And Storage | 4°C |
| Source | Carica papaya latex |
| Quantity | 100 g |
| Product Type | Papain |
| Molecular Weight (g/mol) | 23000 |
| Inhibitors | AEBSF, Antipain, Cystatin, E-64, leupeptin, PMSF, TLCK and TPCK |
| Form | Powder |