missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Papain

Catalog No. ICN10256680
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10256680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10256680 Supplier MP Biomedicals™ Supplier No. 210256680
Only null left

Used in the enzymatic synthesis of amino acids, peptides, and other molecules

Sulfhydryl protease from Carica papaya latex. (A second protease, chymopapain, and a lysozyme have also been isolated from this same source.)

  • Format: Powder
  • Grade: Technical
  • Molecular weight: 23,000
  • Specific Activity:>1750 USP Units/mg

Specifications

Color Off-White
pH 6 to 7
For Use With (Application) Enzymatic synthesis of Amino Acids, Peptides and other molecules
Content And Storage 4°C
Source Carica papaya latex
Quantity 100 g
Product Type Papain
Molecular Weight (g/mol) 23000
Inhibitors AEBSF, Antipain, Cystatin, E-64, leupeptin, PMSF, TLCK and TPCK
Form Powder