missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Nisin

Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)

Supplier:  MP Biomedicals™ 0215583905

 View more versions of this product

Catalog No. ICN15583905

Only null left
Explore available promotions

Description

Description

  • Molecular Formula: C14H230N42O37S7
  • Molecular Weight: 3354.3

Specifications

Specifications

15° to 30°C
Antimicrobial
5 g
Powder
3.31
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions