missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Nisin

Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)

$440.63 - $440.63

Specifications

Content And Storage 15° to 30°C
pH 3.31
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15583905
View Documents
MP Biomedicals™
0215583905
5 g N/A
Only null left
 
Description

Description

  • Molecular Formula: C14H230N42O37S7
  • Molecular Weight: 3354.3

Specifications

Specifications

15° to 30°C
3.31
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'