missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Sodium Iodide (NaI) Solution

Sodium Iodide (NaI) Solution dissolves agarose gels and helps in binding the DNA to the Glassmilk™ binding matrix.

Supplier:  MP Biomedicals™ 111001401

 View more versions of this product

Catalog No. MP111001401


Only null left

Description

Description

  • Less than 15% DNA 1 kb and above is left unbound in NaI.
  • NaI (Sodium Iodide) is supplied with GeneClean™ kits and helps in binding the DNA to the GLASSMILK binding matrix.
  • Melts 0.1 g gel slice in 5 minutes at 55°C.
Specifications

Chemical Identifiers

7681-82-5
15°C to 30°C
Nucleic Acid Isolation and Purification
661°C
NaI
6 M
149.89
NaI
7.0 to 7.4

Specifications

7681-82-5
15°C to 30°C
Nucleic Acid Isolation and Purification
661°C
NaI
6 M
149.89
NaI
7.0 to 7.4
1 x 125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.