missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix I

Catalog No. MP116918100
Change view
Click to view available options
Quantity:
100 x 2 mL Tubes
50 x 2 mL Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116918100 100 x 2 mL Tubes
MP116918050 50 x 2 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116918100 Supplier MP Biomedicals™ Supplier No. 116918100

Lysing Matrix I is effective at breaking very tough and hard samples, including organisms with a tough or dense exterior matrix.

  • Tube Size: 2 mL
  • Sample Type: Plant and animal tissues, wood, seeds, very dense soils and clays, whole insects, ticks, ancient and dried samples, dry grinding fungal spores/rusts
  • Molecule to be Enriched: DNA, RNA, proteins, other small molecules
  • Matrix Composition: Matrix I, contains 2 mm yellow zirconia beads and a 4 mm black ceramic bead

Specifications

Sample Type Seeds, Spores, Animal Tissues, Plants, Soil, Ancient and Dried Samples, Clay, Wood
DNase Dnase-free
For Use With (Application) Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Matrix Lysing matrix I
Quantity 100 x 2 mL Tubes
RNase RNase-free

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.