missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix A, 96-Tube Racks

Lysing Matrix A is used to grind animal and plant tissues, insects, nematodes and lyse bacteria, yeast, fungi, algae. For use on FastPrep™ instruments or any bead beater.

Supplier:  MP Biomedicals™ 116980010

Catalog No. MP116980010


Only null left
Explore available promotions

Description

Description

  • Product Overview: Lysing Matrix A 96-Well Rack comes with 1.2 mL cluster-strip tube and cap assemblies. Each tube is pre-filled with garnet matrix and one 4 mm glass grinding sphere.
  • Applications: Lysing Matrix A is used for all sample types, except soil, for the subsequent isolation of genomic DNA and proteins.
  • Sample Type: Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast, Sludge, Animal Tissue, Bacteria (Gram (+/-), Fixed Samples, Hair, Tooth, Tumor
  • Sample Volume: ≤50 mg per prep
Specifications

Specifications

Garnet Matrix and 4 mm Glass Grinding Sphere
Dnase-free
Nucleic Acid Isolation and Purification, Homogenization
96
RNase-free
Animal Tissues, Plants, Insects, Worms, Bacteria, Yeast, Fungi, Algae
FastPrep-96
Lysing Matrix A
10 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.