missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lipase, USP Grade from Porcine Pancreas

For the resolution of chiral compounds and the transesterification production of biodiesel

$91.93 - $408.17

Specifications

Color Cream
For Use With (Application) Used industrially for the resolution of chiral compounds and the transesterification production of biodiesel
Content And Storage 0°C
Source Porcine pancreas
Product Type Lipase
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10218990
View Documents
MP Biomedicals™
0210218990
500 g
Each for $408.17
Only null left
 
ICN10218980
View Documents
MP Biomedicals™
0210218980
100 g
Each for $91.93
Only null left
 
Description

Description

Lipases are subclass of the esterases. It catalyzes the hydrolysis of emulsified esters of glycerol and long chain fatty acids.

  • Format: Cream powder
  • Partly soluble in water and insoluble in alcohol

Molecular biology

Specifications

Specifications

Cream
0°C
Lipase
Versene, Zn2+, Cu2+, Hg2+, iodine, PCMB
Used industrially for the resolution of chiral compounds and the transesterification production of biodiesel
Porcine pancreas
45000 to 50000
Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'