missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Large Capsules
Contents/liter: LB Medium, 15g Agar-B
$157.88 - $4784.27
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| Format | Capsule |
| Grade | Molecular Biology |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP113002216
|
MP Biomedicals™
113002216 |
227 g |
Each for $157.88
|
|
|||||
|
MP113002226
|
MP Biomedicals™
113002226 |
454 g |
Each for $470.44
|
|
|||||
|
MP113002236
|
MP Biomedicals™
113002236 |
1000 g |
Each for $920.90
|
|
|||||
|
MP113002246
|
MP Biomedicals™
113002246 |
5000 g |
Each for $4,784.27
|
|
|||||
Description
Use 8 large capsules/L.
Specifications
| 15°C to 30°C | |
| Molecular Biology | |
| Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L |
| Capsule | |
| Agar | |
| For growth of bacteria |