missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ LB-Agar, Large Capsules

Contents/liter: LB Medium, 15g Agar-B

Supplier:  MP Biomedicals™ 113002236

 View more versions of this product

Catalog No. MP113002236


Only null left
Explore available promotions

Description

Description

Use 8 large capsules/L.

Specifications

Specifications

Molecular Biology Certified™ FDA
Capsule
Agar
For growth of bacteria
15°C to 30°C
Molecular Biology
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
1000 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions