missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Large Capsules
Contents/liter: LB Medium, 15g Agar-B
Supplier: MP Biomedicals™ 113002246
Description
Use 8 large capsules/L.
Specifications
| Molecular Biology Certified™ | |
| Capsule | |
| Agar | |
| For growth of bacteria |
| 15°C to 30°C | |
| Molecular Biology | |
| Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L | |
| 5000 g |