missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lactase from Aspergillus oryzae

Catalog No. ICN10078005
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10078005 5 g
ICN10078001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10078005 Supplier MP Biomedicals™ Supplier No. 0210078005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleaves lactose into its monosaccharide components, glucose and galactose

  • Hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and α-D-Glucose
  • Catalyses transglycosylation of glucose into allolactose, the inducer of β-galactosidase

  • EC Number: 232-864-1
  • MDL Number: MFCD00130623
  • Format: Off white powder
  • Specific Activity: >5000U/g
  • Stability: Stable in pH range of 4.0 to 8.0
  • Stable to heat
  • Soluble in water, insoluble in ethanol, acetone and isopropyl alcohol
  • Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate

Used to screen for blue white colonies in MCS of various plasmid vectors in Escherichia coli or other bacteria and to break down lactose in milk to make it suitable for people with lactose intolerance.

Specifications

Concentration >5000 U/g
Color Off-White
pH 4 to 8
For Use With (Application) Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk
Content And Storage 4°C
Source Aspergillus oryzae
Quantity 5 g
Product Type Lactase
Form Powder