missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lactase from Aspergillus oryzae

Cleaves lactose into its monosaccharide components, glucose and galactose

Supplier:  MP Biomedicals™ 0210078001

 View more versions of this product

Catalog No. ICN10078001


Only null left

Description

Description

  • Hydrolyzes Lactose (4-o-β-D-Galactopyranosyl-D-glucose) and produces β-D-Galactose and α-D-Glucose
  • Catalyses transglycosylation of glucose into allolactose, the inducer of β-galactosidase

  • EC Number: 232-864-1
  • MDL Number: MFCD00130623
  • Format: Off white powder
  • Specific Activity: >5000U/g
  • Stability: Stable in pH range of 4.0 to 8.0
  • Stable to heat
  • Soluble in water, insoluble in ethanol, acetone and isopropyl alcohol
  • Optimum pH is 4.5 and 4.8 when O-Nitrophenyl-beta-D- galactopyranoside and Lactose are used as substrate

Used to screen for blue white colonies in MCS of various plasmid vectors in Escherichia coli or other bacteria and to break down lactose in milk to make it suitable for people with lactose intolerance.

Specifications

Chemical Identifiers

>5000 U/g
4 to 8
4°C
Lactase
Off-White
Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk
Aspergillus oryzae
Powder

Specifications

>5000 U/g
4 to 8
4°C
1 g
Powder
Off-White
Screen for blue white colonies in MCS of various plasmid vectors in E. coli or other Bacteria and to break down Lactose in milk
Aspergillus oryzae
Lactase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions