missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ L-Glutamic Dehydrogenase
Description
Pyridine nucelotide enzyme which catalyzes the reversible oxidative deamination of L-glutamate to α-keto-glutarate and ammonia. L-glutamic dehydrogenase plays a unique role in mammalian metabolism
- Format: Cloudy white suspension
- Specific Activity: >40 Units/mg
Clinical trials
Specifications
Specifications
| Concentration | 20 mg/mL |
| Color | White |
| pH | 7.6 |
| For Use With (Application) | Diagnosis of liver disease, measurement for the safety of a drug |
| Content And Storage | 4°C |
| Source | Bovine liver |
| Quantity | 500 mg |
| Product Type | L-Glutamic Dehydrogenase |
| Inhibitors | Dimethyl Dicarbonate |
| Form | Suspension |