missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protease (S. Aureus) V.8

Catalog No. MP08399001
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08399001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08399001 Supplier MP Biomedicals™ Supplier No. 08399001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protease (S. Aureus) V.8, also called endoproteinase Glu-C, Glu-C protease, staphylococcal serine proteinase. Widely used for protein sequencing, this enzyme specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids.

  • Protease V.8 specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids
  • In the presence of ammonium, the specificity is limited to glutamic sites
  • Used in a study to investigate immobilized residue-specific endoproteinases for protein sequencing
  • Protease V8 was also used in a study to assess enzymatic cleaning of inorganic ultrafiltration membranes used for whey protein fractionation
  • Provides:
    • Robust activity, greater than 915 units/mg protein
    • Specific cleavage for glutamic acid in ammonium buffer
    • Selective digestion at both glutamic and aspartic acid residues in phosphate buffer at pH 7.8
    • Cleavage activity remains in the presence of many denaturing agent, such as SDS, urea and guanidine-HCl
    • Lyophilized format for longer stability and easier storage

Specifications

For Use With (Application) Used in a study to investigate immobilized residue-specific endoproteinases for Protein sequencing
pH Range 4 and 7.8
Sample Type Staphylococcus aureus
Content And Storage 2°C to 8°C
Form Lyophilized
Quantity 5 mg
Product Type Protease