missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 1X Hanks' Balanced Salt Solution, Modified

Catalog No. ICN1810054
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1810054 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1810054 Supplier MP Biomedicals™ Supplier No. 091810054

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Red, Clear Solution

Used for washing cells and tissues while maintaining cells in a viable state. Buffered with phosphate and maintains a physiological pH. Contains phenol red.

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (HBSS)
Color Red
Concentration 1 X
pH 6.9 to 7.2
Physical Form Liquid
Quantity 500 mL
Grade Not Available

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.