missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Glucose Oxidase
Description
Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.
Specifications
Specifications
| Concentration | >100 U/mg |
| Color | Yellow |
| pH | 4 to 7 |
| For Use With (Application) | Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution |
| Content And Storage | -20°C |
| Source | Aspergillus niger |
| Quantity | 500 kU |
| Product Type | Glucose Oxidase |
| Molecular Weight (g/mol) | ≈160000 |
| Inhibitors | Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose |
| Show More |