missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Glucose Oxidase

Catalog No. ICN19519610
Change view
Click to view available options
Quantity:
10 kU
50 kU
500 kU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19519610 10 kU
ICN19519650 50 kU
ICN19519690 500 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19519610 Supplier MP Biomedicals™ Supplier No. 0219519610

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

From Aspergillus niger

Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.

Specifications

Concentration >100 U/mg
Color Yellow
pH 4 to 7
For Use With (Application) Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Content And Storage -20°C
Source Aspergillus niger
Quantity 10 kU
Product Type Glucose Oxidase
Molecular Weight (g/mol) ≈160000
Inhibitors Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose
Form Lyophilized Powder
Show More Show Less