missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit

Catalog No. MP111102200
Change view
Click to view available options
No. of Reactions:
300 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP111102200 50 Preps
111102600 300 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP111102200 Supplier MP Biomedicals™ Supplier No. 111102200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. MP Biomedicals™ Geneclean™ Turbo Kit can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.

  • Process with either vacuum or centrifugation and save time with patented Turbo filter design
  • Patented GeneClean™ technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute
  • Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications

Specifications

Content And Storage Room Temperature
Format Geneclean Imbedded Turbo Spin Column
Isolation Technology Glass Milk
For Use With (Application) Nucleic Acid isolation and purification
No. of Reactions 50 Preps
Product Type DNA Purification Kit

Research Use Only (RUO)