missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Geneclean™ Turbo Kit
Description
- Process with either vacuum or centrifugation and save time with patented Turbo filter design
- Patented GeneClean™ technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute
- Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications
Specifications
Specifications
| Content And Storage | Room Temperature |
| Format | Geneclean Imbedded Turbo Spin Column |
| For Use With (Application) | Nucleic Acid isolation and purification |
| No. of Reactions | 300 Preps |
Research Use Only (RUO)