missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Spin Kit 200 bp to 300 kb

Used to purify fragments of DNA from 200 bp to 300 kbp from solutions and agarose gels. The GENECLEAN™ SPIN kit uses SPIN filters, which prevent silica particle carry-over into cleaned DNA. Glassmilk™ slurry allows flexibility of reaction size and volume.

$400.52 - $1,764.98

Specifications

CAS 593-84-0
Format Kit
Sample Type Enzymatic Reactions, Solutions, Agarose Gels
For Use With (Application) Isolate DNA from TAE or TBE buffered agarose gels and solutions
Includes SPIN filters
View More Specs

Products 3
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
111101200
View Documents Promotion Details
MP Biomedicals™
111101200
50 Preps
Each for $400.52
Only null left
 
111101400
View Documents Promotion Details
MP Biomedicals™
111101400
100 Preps
Each for $590.48
Only null left
 
111101600
View Documents Promotion Details
MP Biomedicals™
111101600
300 Preps
Each for $1,764.98
Only null left
 
Description

Description

  • The GENECLEAN™ SPIN kit simplifies the process of purifying DNA into three easy steps: Bind, Wash and Elute
  • Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications
  • Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA
Specifications

Specifications

593-84-0
Enzymatic Reactions, Solutions, Agarose Gels
SPIN filters
Spin Kit
DNA Clean-Up Kit
Kit
Isolate DNA from TAE or TBE buffered agarose gels and solutions
Geneclean SPIN New Wash pH Range: 7.4 to 7.7, GC Spin Elution Solution pH Range: 6.8 to 7.2
Up to ∼300 mg Gel Slice or up to ∼300 μL DNA Solution
Gels and Solutions
Videos
SDS
Documents

Documents

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.