missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ for Ancient DNA Kit

Purifies fragmented or damaged DNA from aged or preserved samples

Supplier:  MP Biomedicals™ 111002200

Catalog No. MP111002200


Only null left
Explore available promotions

Includes:
  • Dehybernation Solutions A, A2 and B
  • Ancient DNA GLASSMILK
  • Salton Wash £1 and £2
  • Ancient Alcohol Wash
  • DNA-free Elution Solution
  • Spin filter with tube
  • Catch tube
  • Lysing Matrix K

Description

Description

  • Format: Geneclean™ slurry plus SPIN filter
  • Size: 100 preps

Specifications

Specifications

Geneclean Slurry Plus Spin Filter
Dehybernation solution A, B, A2, Ancient DNA Glassmilk, Ancient DNA Alcohol wash, DNA-free elution solution, Homogenization matrix
Ancient DNA Kit
Purifies fragmented or damaged DNA from aged or preserved samples
100 Preps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions