Learn More
MP Biomedicals™ Screw Caps
These 2 mL Screw Caps are for tubes to be used on FastPrep™ instruments or any bead beater.
$366.25 - $653.85
Specifications
Capacity 2 mL
View More Specs
Products 7
Catalog Number Mfr. No. Quantity Color Price Quantity  
Catalog Number Mfr. No. Quantity Color Price Quantity  
MP115066002
View Documents
MP Biomedicals™
115066002
500 Pk. Red
Each for $509.58
Only null left
MP115063002
View Documents
MP Biomedicals™
115063002
500 Pk. Orange
Each for $366.25
Only null left
115067002
View Documents
MP Biomedicals™
115067002
500 Pk. Green
Each for $366.25
Only null left
115065002
View Documents
MP Biomedicals™
115065002
500 Pk. Blue
Each for $366.25
Only null left
MP115064002
View Documents
MP Biomedicals™
115064002
500 Pk. Purple
Each for $366.25
Only null left
MP115063005
View Documents
MP Biomedicals™
115063005
1000 Pk. Orange
Each for $653.85
Only null left
115065005
View Documents
MP Biomedicals™
115065005
1000 Pk. Blue
Each for $653.85
Only null left
Description
Description
  • Screw Caps with O-rings fit 2 mL screw cap tubes and are ideal for high-speed sample preparation instruments.
  • Screw Caps fit Empty Fastprep™ Tubes and are used in Fastprep systems, to hold sample for Homogenization.
  • Available separately for those who wish to fill their own Lysing Matrix Tubes.
  • Adapter Compatibility: ConeFlex Legacy (allows all FP24 adapters on FP96), CoolPrep (24x2 mL), HiPrep (48x2 mL), QuickFlex (96x2 mL), QuickPrep (24x2 mL), QuickPrep All Metal (24x2 mL)
  • Screw Caps can also be found in the FastDNA and FastDNA SPIN Kits.
Specifications
Specifications
2 mL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.