missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ FastDNA™ SPIN Kit for Feces, 2 mL Tubes
FastDNA™ SPIN Kit for Feces is designed to quickly and efficiently isolates PCR-ready genomic DNA from fresh or frozen human and animal stool samples.
Supplier: MP Biomedicals™ 116570200
Requires : FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin tubes, Microcentrifuge tubes , Sterile tubes for DNA binding, Rotator or low-speed vortex, 100% Ethanol
Description
- Product Overview: The FastDNA SPIN Kit for Feces quickly and efficiently isolates PCR-ready genomic DNA from fresh or frozen human and animal stool samples.
- Host cells, bacteria, fungi, viruses, protists and other cells present in fecal samples are easily lysed within 40 seconds with any FastPrep™ instrument.
- The FastDNA SPIN Kit contains pre-filled 2 mL Lysing Matrix tubes for the homogenization step as well as all the reagents needed to extract DNA in a purification process eliminating PCR inhibitors.
- Sample Type: Feces, Soil, Wastewater, Environmental Water, Sludge
- Sample Volume: Up to 500 mg per prep
Specifications
| Feces, Soil, Wastewater, Environmental Water, Sludge | |
| Use to isolate PCR-ready genomic DNA from fresh or frozen Human and animal stool samples | |
| FastDNA-24 Kits |
| FastPrep™ Instruments | |
| Lysing Matrix E 50 x 2.0 mL tubes, PLS Solution, MT Buffer, Sodium Phosphate Buffer, PPS, Binding Matrix, Wash Buffer #1, Wash Buffer #2, TES, BBS Gel Loading Dye, SPIN Filter Tubes, Catch Tubes | |
| Up to 500 mg per prep |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.