missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Wash Solution SEWS-M, Concentrated

Catalog No. 116540405
Change view
Click to view available options
Quantity:
12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
116540405 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 116540405 Supplier MP Biomedicals™ Supplier No. 116540405
Only null left

Concentrated Wash Solution SEWS-M is used in extraction of DNA to wash away impurities once the DNA is bound to the Binding Matrix.

  • Concentrated SEWS-M is a wash solution included in the FastDNA and all FastDNA Spin Kits and is used to remove impurities once the DNA is bound to the Binding Matrix.
  • Purified DNA is eluted from the Binding Matrix after washing with the SEWS-M solution.
  • Concentrated SEWS-M is an EtOH-based wash solution used in extraction of DNA to remove impurities once the DNA is bound to the Binding Matrix.

Specifications

Sample Type Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1
For Use With (Application) Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
Product Type Concentrated SEWS-M
Quantity 12 mL
Sample Volume Up to 500 mg Per Prep