missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Collagenase from Clostridium histolyticum
Enzyme responsible for break down of peptide bonds in collagen.
Supplier: MP Biomedicals™ 021951091
Description
Assist in destroying extracellular structures in pathogenesis of bacteria such as Clostridium. These enzymes are exotoxins (a virulence factor) and help to facilitate the spread of gas gangrene. They normally target the connective tissue in muscle cells and other body organs.
- Synonyms: Clostridiopeptidase A
- CAS Number: 9001-12-1
- EC Number: 232-582-9
- MDL Number: MFCD00130830
- Format: Dialyzed, lyophilized brown lumpy powder
- Specific Activity: >125U/mg
- Solubility: Dissolve enzyme at concentration of 1mg/mL in 0.05M TES with 0.36mM calcium chloride, pH 7.5
Note: This item is for research use only. It is not a compendial item and is not meant for medical use.
Specifications
| >125 U/mg | |
| 7 to 9 | |
| Clostridium histolyticum | |
| Collagenase | |
| Metal chelating agents such as Cysteine, EDTA or o-Phenanthroline |
| Brown | |
| 4°C | |
| 100 mg | |
| 105000 and 57400 | |
| Lyophilized Powder |