missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Cholesterol Esterase from Pseudomonas Species
Description
- CAS Number: 9026-00-0
- EC Number: 232-808-6
- MDL Number: MFCD00071052
- Format: yellowish tan lyophilized powder
- Specific activity: ≥100U/mg (solid)
- Isoelectric point: 5.9 ±0.1
- Stable at -20°C for at least one year
Cholesterol Esterase is useful for enzymatic determination of total cholesterol when coupled with cholesterol oxidase in clinical analysis.
Specifications
Specifications
| Concentration | ≥100 U/mg |
| Color | Yellowish Tan |
| pH | 7 to 9 |
| For Use With (Application) | Enzymatic determination of Total Cholesterol when coupled with Cholesterol Oxidase in clinical analysis |
| Content And Storage | 0°C |
| Source | Pseudomonas species |
| Quantity | 200 U |
| Product Type | Cholesterol Esterase |
| Molecular Weight (g/mol) | ≈300000 |
| Inhibitors | Hg- - Ag ionic detergents |
| Show More |