Learn More
MP Biomedicals™ Cellulase from Aspergillus niger
Cellulase from Aspergillus niger catalyzes hydrolysis of endo-1,4-β-D-glycosidic linkages in cellulose, lichenin, barley glucan and cellooligosaccharides cellotriose to cellohexaose
Supplier: MP Biomedicals™ 0215058380
Description
Cellulase refers to a group of enzymes which acting together hydrolyze cellulose. The b-glucosidases, including cellobiose are primarily active on the smaller molecular weight cellulose hydrolysates.
- CAS Number: 9012-54-8
- EC Number: 232-734-4
- MDL Number: MFCD00081510
- Format: Tan powder
- Specific activity: >60000U/g
- Readily soluble in water
- Foreign activity: Salmonella Negative
- Optimum pH: 4.2 to 5.2
Cellulase is used in molecular biology, cell biology, textile industry, pulp and paper industry and for commercial food processing in coffee
Chemical Identifiers
| >60,000 U/g | |
| 4.2 to 5.2 | |
| 4°C | |
| Cellulase |
| Tan | |
| Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels | |
| Aspergillis niger | |
| Powder |
Specifications
| >60,000 U/g | |
| 4.2 to 5.2 | |
| 4°C | |
| 100 g | |
| Powder |
| Tan | |
| Cellulase is used for commercial food processing in coffee, textile industry and in laundry detergents, pharmaceutical applications, fermentation of biomass into biofuels | |
| Aspergillis niger | |
| Cellulase |