missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Catalase from Beef Liver

Catalog No. icn10042801
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10042801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10042801 Supplier MP Biomedicals™ Supplier No. 0210042801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxidoreductase enzyme for catalyzing conversion of hydrogen peroxide to water and oxygen, 2H2O2 → 2H2O + O2

Hydrogen peroxide is a harmful byproduct of many normal metabolic processes. to prevent damage to cells and tissues, it must be quickly converted into other, less dangerous substances. To this end, catalase is frequently used by cells to rapidly catalyze the decomposition of hydrogen peroxide into less-reactive gaseous oxygen and water molecules.

  • CAS Number: 9001-05-2
  • EC Number: 232-577-1
  • MDL Number: MFCD00081483
  • Format: Dark green powder
  • Specific Activity: ∽13,000U/mg solid
  • Extinction Coefficient: 36.5 (276nm)
  • Isoelectric point (pI): 5.4
  • Dissolves readily at 5mg/mL in 0.05M sodium/potassium phosphate (pH 7.0) giving clear green solution

Used in the food industry for removing hydrogen peroxide from milk prior to cheese production.

Specifications

Concentration ≈13,000 U/mg
Color Dark Green
For Use With (Application) Used in the food industry for removing Hydrogen Peroxide from milk prior to cheese production
Content And Storage 0°C
Source Beef liver
Quantity 1 g
Product Type Catalase
Molecular Weight (g/mol) 250
Form Lyophilized Powder