missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein sodium salt

Casein sodium salt White to off-white powder

$77.24 - $582.00

Specifications

Form Powder
Color White
Synonym Sodium Caseinate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN90289601
View Documents
MP Biomedicals™
0290289601
1 kg
Each for $145.76
Only null left
 
ICN90289605
View Documents
MP Biomedicals™
0290289605
5 kg
Each for $582.00
Only null left
 
MP29028965
View Documents
MP Biomedicals™
029028965
500 g
Each for $77.24
Only null left
 
Description

Description

  • Casein protein has numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
  • Casein sodium salt from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk: α-s1 Casein, α-s2 Casein, β-Casein, and κ-Casein.
  • Casein is proposed to be the main protective constituent in milk.
Specifications

Specifications

Powder
White
Sodium Caseinate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'