missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casein sodium salt

Casein sodium salt White to off-white powder

Supplier:  MP Biomedicals™ 0290289601

 View more versions of this product

Catalog No. ICN90289601


Only null left
Explore available promotions

Description

Description

  • Casein protein has numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
  • Casein sodium salt from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk: α-s1 Casein, α-s2 Casein, β-Casein, and κ-Casein.
  • Casein is proposed to be the main protective constituent in milk.
Specifications

Specifications

Casein Sodium salt
1 kg
Powder
Soluble in water (1% aq soln-Cloudy Solution).
6.5 to 7.5
15° to 30°C
White
Sodium Caseinate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions