missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Carbonic Anhydrase

Catalog No. ICN15387980
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15387980 100 mg
ICN15387925 25 mg
ICN15387910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15387980 Supplier MP Biomedicals™ Supplier No. 0215387980

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Activity 2000units/mg

Specifications

Concentration >2000 U/mg
Color Tan
For Use With (Application) Molecular biology, cell biology
Content And Storage 2°C to 8°C
Source Bovine erythrocytes
Quantity 100 mg
Product Type Carbonic Anhydrase
Molecular Weight (g/mol) 30000
Form Lyophilized Powder