missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Apoferritin from Horse Spleen

Protein shell of ferritin, abundant in beta-cells of pancreas

$188.94 - $1738.40

Specifications

For Use With (Application) Molecular Biology
Molecular Weight (g/mol) 445000
Name Apoferritin from Horse Spleen
Source Horse Spleen
Storage Requirements -20°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10026080
View Documents
MP Biomedicals™
0210026080
100 mg
Each for $536.99
Only null left
 
ICN10026025
View Documents
MP Biomedicals™
0210026025
25 mg
Each for $188.94
Only null left
 
ICN10026090
View Documents
MP Biomedicals™
0210026090
500 mg
Each for $1,738.40
Only null left
 
Description

Description

When added to cultured endothelial cells apoferritin is taken up in a dose-responsive manner and appears to protect the cells from oxidant-mediated cytolysis.

  • CAS Number: 9013-31-4
  • MDL Number: MFCD00081365
  • Format: Off-white lyophilized powder
  • Protein Content: ≈90%
  • Iron Content: Typically <0.01%
  • Soluble in water or dilute buffer

Used for calibration of gel filtration columns and sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis. It acts as an anti-oxidant.

Specifications

Specifications

Molecular Biology
Apoferritin from Horse Spleen
-20°C
445000
Horse Spleen
Lyophilized
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'